3-Fluoro-(4-methoxycarbonyl)phenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(3-Fluoro-4-(methoxycarbonyl)phenyl)boronic acid, 3-Fluoro-4-(methoxycarbonyl)phenylboronic acid, 3-Fluoro-4-methoxycarbonylphenylboronic acid, 3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid
H52510-03
H52510-06
AAH52510-03EA
109
USD
AAH52510-03
AAH52510-06
AAH52510-MD
3-Fluoro-(4-methoxycarbonyl)phenylboronic acid 97%
3-Fluoro-(4-methoxycarbonyl)phenylboronic acid
Purity: 97%
CAS No.: 505083-04-5
Molecular Formula: C8H8BFO4
Formula Weight: 197.96
Melting Point: 180-184°
MDL No.: MFCD04115665
Formula:
C₈H₈BFO₄ MW: 197.96 g/mol Melting Pt: 180…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115665 CAS Number: 505083-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...