4-Fluoro-3-(hydroxymethyl)benzeneboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Fluoro-3-(hydroxymethyl)phenylboronic acid, 4-Fluoro-3-(hydroxymethyl)benzeneboronic acid

H52487-03 H52487-MD
AAH52487-03EA 239 USD
AAH52487-03 AAH52487-MD
4-Fluoro-3-(hydroxymethyl)benzeneboronic acid 98%
4-Fluoro-3-(hydroxymethyl)benzeneboronic acid

Purity: 98% CAS No.: 481681-02-1 Molecular Formula: C7H8BFO3 Formula Weight: 169.95 Melting Point: 124-126° MDL No.: MFCD08235075

Formula: C₇H₈BFO₃
MW: 169.95 g/mol
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD08235075
CAS Number: 481681-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR