2-(Bromomethyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Bromomethyl)benzeneboronic acid, Dihydroxy(2-(bromomethyl)phenyl)borane, (2-(Bromomethyl)phenyl)metaboric acid, 2-Bromomethylphenylboronic acid

H52467-03 H52467-06
AAH52467-03EA 80.5 USD
AAH52467-03 AAH52467-06 AAH52467-MD
2-(Bromomethyl)phenylboronic acid 97%
2-(Bromomethyl)phenylboronic acid

Purity: 97% CAS No.: 91983-14-1 Molecular Formula: C7H8BBrO2 Formula Weight: 214.85 Melting Point: 145-148° MDL No.: MFCD01318941

Formula: C₇H₈BBrO₂
MW: 214.85 g/mol
Melting Pt: 145…148 °C
Storage Temperature: Ambient
MDL Number: MFCD01318941
CAS Number: 91983-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR