3-(tert-Butoxycarbonyl)aminomethylphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-(((tert-Butoxycarbonyl)amino)methyl)phenyl)boronic acid, 3-(Aminomethyl)benzeneboronic acid, N-BOC protected

H52453-03 H52453-MD
AAH52453-03EA 134 USD
AAH52453-03 AAH52453-MD
3-(tert-Butoxycarbonyl)aminomethylphenylboronic acid 97%
3-(tert-Butoxycarbonyl)aminomethylphenylboronic acid

Purity: 97% CAS No.: 199609-62-6 Molecular Formula: C12H18BNO4 Formula Weight: 251.09 Melting Point: 188-190° MDL No.: MFCD06246052

Formula: C₁₂H₁₈BNO₄
MW: 251.09 g/mol
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD06246052
CAS Number: 199609-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR