Boc-Lys(ME)2-OH 97%
Supplier: Thermo Scientific Chemicals
H52437-03
H52437-MD
AAH52437-03EA
233
USD
AAH52437-03
AAH52437-MD
Boc-Lys(ME)2-OH 97%
Boc-Lys(ME)2-OH
Purity: 97%
CAS No.: 65671-53-6
Molecular Formula: C13H26N2O4
Formula Weight: 274.36
MDL No.: MFCD00076965
Formula:
C₁₃H₂₆N₂O₄ MW: 274.361 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00076965 CAS Number: 65671-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...