Boc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-Val(3-OH)-OH, 3-Hydroxy-L-valine, N-BOC protected, (S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxy-3-methylbutanoic acid

H52429-03 H52429-MD
AAH52429-03EA 937 USD
AAH52429-03 AAH52429-MD
Boc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid 97%
Boc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid

Purity: 97% CAS No.: 102507-13-1 Molecular Formula: C10H19NO5 Formula Weight: 233.26 MDL No.: MFCD03094792

Formula: C₁₀H₁₉NO₅
MW: 233.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094792
CAS Number: 102507-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR