D-erythro-Sphingosine hydrochloride 97%

Supplier: Thermo Scientific Chemicals

H52427-MC H52427-ME
AAH52427-MCEA 284 USD
AAH52427-MC AAH52427-ME
D-erythro-Sphingosine hydrochloride 97%
D-erythro-Sphingosine hydrochloride

Inhibitor of protein kinase C and calmodulin-dependent enzymes, but may stimulate mast cells by activation of protein kinase C


Purity: 97% CAS No.: 2673-72-5 Molecular Formula: C18H37NO2.HCl Formula Weight: 335.96 MDL No.: MFCD08436977

Formula: C₁₈H₃₇NO₂·HCl
MW: 335.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08436977
CAS Number: 2673-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR