(R)-3-Amino-1-benzylpiperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

H52425-03 H52425-06
AAH52425-03EA 84.1 USD
AAH52425-03 AAH52425-06
(R)-3-Amino-1-benzylpiperidine 97%
(R)-3-Amino-1-benzylpiperidine

Purity: 97% CAS No.: 168466-84-0 Molecular Formula: C12H18N2 Formula Weight: 190.29 MDL No.: MFCD03093374

Formula: C₁₂H₁₈N₂
Storage Temperature: Ambient
MDL Number: MFCD03093374
CAS Number: 168466-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR