N-Boc-2-methyl-D-serine methyl ester 97%

Supplier: Thermo Scientific Chemicals

H52421-03 H52421-MD
AAH52421-03EA 491 USD
AAH52421-03 AAH52421-MD
N-Boc-2-methyl-D-serine methyl ester 97%
N-Boc-2-methyl-D-serine methyl ester

Purity: 97% CAS No.: 188476-33-7 Molecular Formula: C10H19NO5 Formula Weight: 233.26 MDL No.: MFCD06797553

Formula: C₁₀H₁₉NO₅
MW: 233.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797553
CAS Number: 188476-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR