(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
tert-Butyl (R)-2-(Aminomethyl)-1-pyrrolidinecarboxylate, (R)-2-(Aminomethyl)-1-Boc-pyrrolidine, (R)-2-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine, (R)-2-(Aminomethyl)-1-pyrrolidinecarboxylic acid tert-Butyl ester
H52419-03
H52419-06
H52419-MD
AAH52419-03EA
86.9
USD
AAH52419-03
AAH52419-06
AAH52419-MD
(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine 97%
(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine
Purity: 97%
CAS No.: 259537-92-3
Molecular Formula: C10H20N2O2
Formula Weight: 200.28
Boiling Point: 98-110°/1mm
Optical Rotation: +47° (c=1 in chloroform)
UN No.: 2735
MDL No.: MFCD03419256
Formula:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol Boiling Pt: 98…110 °C/1 mm Storage Temperature: Ambient |
MDL Number:
MFCD03419256 CAS Number: 259537-92-3 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...