(1R,2R)-2-Aminocyclohexanecarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H52418-03
H52418-MD
AAH52418-03EA
605
USD
AAH52418-03
AAH52418-MD
(1R,2R)-2-Aminocyclohexanecarboxylic acid 97%
(1R,2R)-2-Aminocyclohexanecarboxylic acid
Purity: 97%
CAS No.: 26685-83-6
Molecular Formula: C7H13NO2
Formula Weight: 143.19
Optical Rotation: -77° (c=1 in water)
MDL No.: MFCD08276263
Formula:
C₇H₁₃NO₂ MW: 143.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08276263 CAS Number: 26685-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...