(R)-1-Boc-piperidine-3-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid, Boc-D-Nip-OH, 1-Boc-D-nipecotic acid, (R)-N-BOC-piperidine-3-carboxylic acid
H52416-06
H52416-14
AAH52416-06EA
101
USD
AAH52416-06
AAH52416-14
(R)-1-Boc-piperidine-3-carboxylic acid 97%
(R)-1-Boc-piperidine-3-carboxylic acid
Purity: 97%
CAS No.: 163438-09-3
Molecular Formula: C11H19NO4
Formula Weight: 229.28
Beilstein Reference: 5539297
MDL No.: MFCD00673775
Formula:
C₁₁H₁₉NO₄ MW: 229.28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02179173 CAS Number: 163438-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...