3-Boc-aminopyrrolidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(BOC-amino)pyrrolidine, 2-Methyl-2-propanyl 3-pyrrolidinylcarbamate, 1,1-Dimethylethyl3-pyrrolidinylcarbamate, 3-Boc-Aminopyrrolidine

H52414-03 H52414-06
AAH52414-03EA 51.6 USD
AAH52414-03 AAH52414-06
3-Boc-aminopyrrolidine 97%
3-Boc-aminopyrrolidine

Purity: 97% CAS No.: 99724-19-3 Molecular Formula: C9H18N2O2 Formula Weight: 186.25 Melting Point: 54° MDL No.: MFCD00059040

Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Melting Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00059040
CAS Number: 99724-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR