(S)-1-Boc-2-isobutylpiperazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H52409-03 H52409-MD
AAH52409-03EA 424 USD
AAH52409-03 AAH52409-MD
(S)-1-Boc-2-isobutylpiperazine 97%
(S)-1-Boc-2-isobutylpiperazine

Purity: 97% CAS No.: 674792-06-4 Molecular Formula: C13H26N2O2 Formula Weight: 242.36 MDL No.: MFCD03787916

Formula: C₁₃H₂₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03787916
CAS Number: 674792-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR