2-Bromo-1-methylimidazole 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromo-1-methyl-1H-imidazole

H52407-03 H52407-06
AAH52407-03EA 118 USD
AAH52407-03 AAH52407-06
2-Bromo-1-methylimidazole 95%
2-Bromo-1-methylimidazole

Purity: 95% CAS No.: 16681-59-7 Molecular Formula: C4H5BrN2 Formula Weight: 161.00 Flash Point: 110°(230°F) Boiling Point: 172° MDL No.: MFCD02179525

Formula: C₄H₅BrN₂
MW: 161 g/mol
Boiling Pt: 172 °C
Density: 1.649
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD02179525
CAS Number: 16681-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR