Ethyl-4-methyloxazole-5-carboxylate ≥97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-methyloxazole-5-carboxylate

H52404-03 H52404-06
AAH52404-03EA 142 USD
AAH52404-03 AAH52404-06 AAH52404-MD
Ethyl-4-methyloxazole-5-carboxylate ≥97%
Ethyl-4-methyloxazole-5-carboxylate

Purity: 97+% CAS No.: 20485-39-6 Molecular Formula: C6H7NO3 Formula Weight: 141.10 Physical Form: Liquid Appearance: Colorless MDL No.: MFCD00062573

Formula: C₇H₉NO₃
MW: 155.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00062573
CAS Number: 20485-39-6
EINECS: 243-848-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR