1-(2-Bromoethoxy)-4-fluorobenzene 97+%

Supplier: Thermo Scientific Chemicals
Warning

H52403-03 H52403-06
AAH52403-03EA 107 USD
AAH52403-03 AAH52403-06 AAH52403-MD
1-(2-Bromoethoxy)-4-fluorobenzene 97+%
1-(2-Bromoethoxy)-4-fluorobenzene

Purity: 97+% CAS No.: 332-48-9 Molecular Formula: C8H8BrFO Formula Weight: 219.05 MDL No.: MFCD00044739

Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD00044739
CAS Number: 332-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR