4-Methoxyindole-2-carboxylic acid ≥97%

Supplier: Thermo Scientific Chemicals

H52392-03 H52392-06 H52392-MD
AAH52392-03EA 146 USD
AAH52392-03 AAH52392-06 AAH52392-MD
4-Methoxyindole-2-carboxylic acid ≥97%
4-Methoxyindole-2-carboxylic acid

Purity: 97+% CAS No.: 103260-65-7 Molecular Formula: C10H9NO3 Formula Weight: 191.19 MDL No.: MFCD02664458

Formula: C₁₀H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD02664458
CAS Number: 103260-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR