7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥97%

Supplier: Thermo Scientific Chemicals
Warning

H52389-03 H52389-06 H52389-MD
AAH52389-03EA 187 USD
AAH52389-03 AAH52389-06 AAH52389-MD
7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥97%
7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride

Purity: 97+% CAS No.: 99365-69-2 Molecular Formula: C9H10N2O2.HCl Formula Weight: 214.65 MDL No.: MFCD07371494

Formula: C₉H₁₁ClN₂O₂
MW: 214.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371494
CAS Number: 99365-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR