1-(2-Aminoethyl)-4-methylpiperazine 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-(4-Methylpiperazin-1-yl)ethanamine, 2-(4-Methyl-piperazin-1-yl)-ethylamine
H52371-03
H52371-06
H52371-MD
AAH52371-03EA
146
USD
AAH52371-03
AAH52371-06
AAH52371-MD
1-(2-Aminoethyl)-4-methylpiperazine 97+%
1-(2-Aminoethyl)-4-methylpiperazine
Purity: 97+%
CAS No.: 934-98-5
Molecular Formula: C7H17N3
Formula Weight: 143.43
UN No.: 2735
MDL No.: MFCD03701701
Formula:
C₇H₁₇N₃ MW: 143.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03701701 CAS Number: 934-98-5 EINECS: 213-296-3 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...