2-Fluoro-5-methoxybenzoic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

H52365-03 H52365-06 H52365-MD
AAH52365-03EA 164 USD
AAH52365-03 AAH52365-06 AAH52365-MD
2-Fluoro-5-methoxybenzoic acid 97+%
2-Fluoro-5-methoxybenzoic acid

Purity: 97+% CAS No.: 367-83-9 Molecular Formula: C8H7FO3 Formula Weight: 170.14 MDL No.: MFCD00272566

Formula: C₈H₇FO₃
MW: 170.14
Storage Temperature: Ambient
MDL Number: MFCD00272566
CAS Number: 367-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR