4-Amino-3-bromo-2-chloropyridine 97+%

Supplier: Thermo Scientific Chemicals
Warning

H52359-03 H52359-06
AAH52359-03EA 146 USD
AAH52359-03 AAH52359-06 AAH52359-MD
4-Amino-3-bromo-2-chloropyridine 97+%

Purity: 97+% CAS No.: 215364-85-5 Molecular Formula: C5H4BrClN2 Formula Weight: 207.46 MDL No.: MFCD09263478

Formula: C₅H₄BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD09263478
CAS Number: 215364-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR