4-Amino-3-fluorophenylboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-3-fluorobenzeneboronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Amino-3-fluorophenylboronic acid, pinacol ester

H52349-03 H52349-MD
AAH52349-03EA 89.1 USD
AAH52349-03 AAH52349-MD
4-Amino-3-fluorophenylboronic acid pinacol ester 96%
4-Amino-3-fluorophenylboronic acid pinacol ester

Purity: 96% CAS No.: 819058-34-9 Molecular Formula: C12H17BFNO2 Formula Weight: 237.08 MDL No.: MFCD09033884

Formula: C₁₂H₁₇BFNO₂
MW: 237.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09033884
CAS Number: 819058-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR