N-(2-Hydroxyethyl)formamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H52345-03 H52345-MD
AAH52345-03EA 339 USD
AAH52345-03 AAH52345-MD
N-(2-Hydroxyethyl)formamide 97%
N-(2-Hydroxyethyl)formamide

Purity: 97% CAS No.: 693-06-1 Molecular Formula: C3H7NO2 Formula Weight: 89.09 MDL No.: MFCD00021040

Formula: C₃H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00021040
CAS Number: 693-06-1
EINECS: 211-741-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR