2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylate

H52312-03 H52312-MD
AAH52312-03EA 258 USD
AAH52312-03 AAH52312-MD
2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester 96%
2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester

Purity: 96% CAS No.: 1073338-92-7 Molecular Formula: C13H19BO5 Formula Weight: 266.10

Formula: C₁₃H₁₉BO₅
Storage Temperature: Ambient
MDL Number: MFCD11855980
CAS Number: 1073338-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR