2-Bromo-3-(bromomethyl)pyridine 96%

Supplier: Thermo Scientific Chemicals
Warning

H52311-03 H52311-MD
AAH52311-03EA 275 USD
AAH52311-03 AAH52311-MD
2-Bromo-3-(bromomethyl)pyridine 96%
2-Bromo-3-(bromomethyl)pyridine

Purity: 96% CAS No.: 94446-97-6 Molecular Formula: C6H5Br2N Formula Weight: 250.92 Flash Point: 110°(230°F) Boiling Point: 291° MDL No.: MFCD08276299

Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Boiling Pt: 291 °C
Melting Pt: 38 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD08276299
CAS Number: 94446-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR