N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine ≥98%

Supplier: Thermo Scientific Chemicals

H52309-03 H52309-06
AAH52309-03EA 177 USD
AAH52309-03 AAH52309-06
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine ≥98%
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine

Purity: 98+% CAS No.: 51549-39-4 Molecular Formula: C23H31N5O4Si Formula Weight: 469.62 MDL No.: MFCD04972281

Formula: C₂₃H₃₁N₅O₄Si
MW: 469.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972281
CAS Number: 51549-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR