Boc-5-amino-2-chlorobenzoic acid ≥98%

Supplier: Thermo Scientific Chemicals
Warning

H52308-03 H52308-06
AAH52308-03EA 188 USD
AAH52308-03 AAH52308-06 AAH52308-MD
Boc-5-amino-2-chlorobenzoic acid ≥98%
Boc-5-amino-2-chlorobenzoic acid

Purity: 98+% CAS No.: 503555-96-2 Molecular Formula: C12H14ClNO4 Formula Weight: 271.70 MDL No.: MFCD02682166

Formula: C₁₂H₁₄ClNO₄
Storage Temperature: Ambient
MDL Number: MFCD02682166
CAS Number: 503555-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR