N-Benzoyl-3'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine ≥97%

Supplier: Thermo Scientific Chemicals

H52306-03 H52306-06
AAH52306-03EA 206 USD
AAH52306-03 AAH52306-06 AAH52306-MD
N-Benzoyl-3'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine ≥97%
N-Benzoyl-3'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine

Purity: 97+% CAS No.: 140712-79-4 Molecular Formula: C38H35N5O6 Formula Weight: 657.73 MDL No.: MFCD04972282

Formula: C₃₈H₃₅N₅O₆
MW: 657.73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972282
CAS Number: 140712-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR