Octahydro-2H-pyrido[1,2-a]pyrazine ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,4-Diazabicyclo[4.4.0]decane
H52305-03
H52305-MD
AAH52305-03EA
499
USD
AAH52305-03
AAH52305-MD
Octahydro-2H-pyrido[1,2-a]pyrazine ≥98%
Octahydro-2H-pyrido[1,2-a]pyrazine
Purity: 98+%
CAS No.: 4430-75-5
Molecular Formula: C8H16N2
Formula Weight: 140.23
Boiling Point: 68-72°/3mm
UN No.: 3267
MDL No.: MFCD01098257
Formula:
C₈H₁₆N₂ MW: 140.23 g/mol Boiling Pt: 68…72 °C/3 mm Storage Temperature: Ambient |
MDL Number:
MFCD01098257 CAS Number: 4430-75-5 UN: 3267 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...