(S)-(-)-2-Amino-3-benzyloxy-1-propanol ≥98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: H-D-Ser(Bzl)-ol

H52304-03 H52304-06
AAH52304-03EA 94.8 USD
AAH52304-03 AAH52304-06 AAH52304-MD
(S)-(-)-2-Amino-3-benzyloxy-1-propanol ≥98%
(S)-(-)-2-Amino-3-benzyloxy-1-propanol

Purity: 98+% CAS No.: 58577-88-1 Molecular Formula: C10H15NO2 Formula Weight: 181.23 UN No.: 3267 MDL No.: MFCD04112477

Formula: C₁₀H₁₅NO₂
MW: 181.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04112477
CAS Number: 58577-88-1
UN: 3267
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR