N-Benzoyl-2'-deoxyadenosine 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: N-(9-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide, N6-Benzoyl-2'-deoxyadenosine, N-Benzoyl-2'-deoxy-adenosine

H52303-03 H52303-06
AAH52303-03EA 114 USD
AAH52303-03 AAH52303-06 AAH52303-MD
N-Benzoyl-2'-deoxyadenosine 98+%
N-Benzoyl-2'-deoxyadenosine

Reactant in the synthesis of 2', 5'-Dideoxycytidines and Other Derivatives of 2'-Deoxycytidine. Reactant in the synthesis of Mono- and Diamino Analogues of 2?-Deoxyadenosine, Cordycepin, 9-(3-Deoxy-α-D-Threo-Pentofuranosyl)-Adenine and 9-(2-Deoxy-α-D-Threo-Pentofuranosyl)Adenine.


Purity: 98+% CAS No.: 4546-72-9 Molecular Formula: C17H17N5O4 Formula Weight: 355.35 MDL No.: MFCD00009628

Formula: C₁₇H₁₇N₅O₄
MW: 355.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009628
CAS Number: 4546-72-9
EINECS: 224-903-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR