5'-O-(tert-Butyldimethylsilyl)thymidine ≥97%

Supplier: Thermo Scientific Chemicals

H52301-03 H52301-MD
AAH52301-03EA 155.36 USD
AAH52301-03 AAH52301-MD
5'-O-(tert-Butyldimethylsilyl)thymidine ≥97%
5'-O-(tert-Butyldimethylsilyl)thymidine

Purity: 97+% CAS No.: 40733-28-6 Molecular Formula: C16H28N2O5Si Formula Weight: 356.49 MDL No.: MFCD01631041

Formula: C₁₆H₂₈N₂O₅Si
MW: 356.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631041
CAS Number: 40733-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR