(S)-2-Amino-3-benzyloxy-1-propanol hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-2-Amino-3-(benzyloxy)propan-1-ol hydrochloride

H52297-03 H52297-06
AAH52297-03EA 83.5 USD
AAH52297-03 AAH52297-06 AAH52297-MD
(S)-2-Amino-3-benzyloxy-1-propanol hydrochloride ≥98%
(S)-2-Amino-3-benzyloxy-1-propanol hydrochloride

Purity: 98+% CAS No.: 61366-43-6 Molecular Formula: C10H15NO2.HCl Formula Weight: 217.70 MDL No.: MFCD07368914

Formula: C₁₀H₁₅NO₂·HCl
MW: 217.70 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368914
CAS Number: 61366-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR