3-Azetidinecarboxylic acid 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Azetanecarboxylic acid, Azetidine-3-carboxylic acid
H52294-03
H52294-06
H52294-MD
AAH52294-03EA
294
USD
AAH52294-03
AAH52294-06
AAH52294-MD
3-Azetidinecarboxylic acid 98+%
3-Azetidinecarboxylic acid
Purity: 98+%
CAS No.: 36476-78-5
Molecular Formula: C4H7NO2
Formula Weight: 101.11
Melting Point: ca 250° dec.
MDL No.: MFCD00191763
Formula:
C₄H₇NO₂ MW: 101.11 g/mol Melting Pt: ca 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00191763 CAS Number: 36476-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...