(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(1R,2R)-Cyclohexane-1,2-dicarboxylic acid, (1R,2R)-(-)-trans-Cyclohexane-1,2-dicarboxylic acid
H52293-03
H52293-06
H52293-MD
AAH52293-03EA
80.8
USD
AAH52293-03
AAH52293-06
AAH52293-MD
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid 98+%
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid
Purity: 98+%
CAS No.: 46022-05-3
Molecular Formula: C8H12O4
Formula Weight: 172.18
Physical Form: Solid
Appearance: White
MDL No.: MFCD00062973
Formula:
C₈H₁₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00062973 CAS Number: 46022-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...