(1S,2S)-(+)-1,2-Cyclohexanedicarboxylic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(1S,2S)-Cyclohexane-1,2-dicarboxylic acid
H52290-03
H52290-06
AAH52290-03EA
89.5
USD
AAH52290-03
AAH52290-06
AAH52290-MD
(1S,2S)-(+)-1,2-Cyclohexanedicarboxylic acid 98+%
(1S,2S)-(+)-1,2-Cyclohexanedicarboxylic acid
Purity: 98+%
CAS No.: 21963-41-7
Molecular Formula: C8H12O4
Formula Weight: 172.18
MDL No.: MFCD00151258
Formula:
C₈H₁₂O₄ MW: 172.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00151258 CAS Number: 21963-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...