(2-Aminoethoxy)acetic acid 98%
Supplier: Thermo Scientific Chemicals
H52289-03
H52289-06
AAH52289-03EA
189
USD
AAH52289-03
AAH52289-06
(2-Aminoethoxy)acetic acid 98%
(2-Aminoethoxy)acetic acid
Purity: 98%
CAS No.: 10366-71-9
Molecular Formula: C4H9NO3
Formula Weight: 119.12
Melting Point: 177-179°
MDL No.: MFCD09865417
Formula:
C₄H₉NO₃ MW: 119.12 g/mol Melting Pt: 177…179 °C Storage Temperature: Ambient |
MDL Number:
MFCD09865417 CAS Number: 10366-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...