Hafnium(IV) isopropoxide isopropanol adduct 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hafnium(IV) i-propoxide monoisopropylate

H52288-14 H52288-22
AAH52288-14EA 386 USD
AAH52288-14 AAH52288-22
Hafnium(IV) isopropoxide isopropanol adduct 99%
Hafnium(IV) isopropoxide isopropanol adduct

Purity: 99% CAS No.: 2171-99-5 Molecular Formula: C12H28HfO4.C3H8O Formula Weight: 474.94 UN No.: 3181 MDL No.: MFCD00070460

Formula: C₁₂H₂₈HfO₄·C₃H₈O
MW: 474.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070460
CAS Number: 2171-99-5
UN: 3181
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR