5-Bromo-1,3-dimethyl-2-pyridone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H52279-03
H52279-06
AAH52279-03EA
127
USD
AAH52279-03
AAH52279-06
5-Bromo-1,3-dimethyl-2-pyridone 97%
5-Bromo-1,3-dimethyl-2-pyridone
Purity: 97%
CAS No.: 51417-13-1
Molecular Formula: C7H8BrNO
Formula Weight: 202.05
Melting Point: 105-106°C
MDL No.: MFCD16658881
Formula:
C₇H₈BrNO Melting Pt: 105…106°C Storage Temperature: Ambient |
MDL Number:
MFCD16658881 CAS Number: 51417-13-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...