Fmoc-O-t-butyl-L-β-homotyrosine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-β-HoTyr(tBu)-OH, Fmoc-β-HomoTyr(tBu)-OH
H52192-03
H52192-MD
AAH52192-03EA
405
USD
AAH52192-03
AAH52192-MD
Fmoc-O-t-butyl-L-β-homotyrosine 95%
Fmoc-O-t-butyl-L-beta-homotyrosine
Purity: 95%
CAS No.: 219967-69-8
Molecular Formula: C29H31NO5
Formula Weight: 473.57
Beilstein Reference: 8236890
MDL No.: MFCD01862862
Formula:
C₂₉H₃₁NO₅ MW: 473.57 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01862862 CAS Number: 219967-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...