Boc-L-beta-homoaspartic acid gamma-benzyl ester 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-β-HomoAsp(OBzl)-OH, Boc-β-HoAsp(OBzl)-OH

H52191-03 H52191-MD
AAH52191-03EA 410 USD
AAH52191-03 AAH52191-MD
Boc-L-beta-homoaspartic acid gamma-benzyl ester 95%
Boc-L-beta-homoaspartic acid-gamma-benzyl ester

Purity: 95% CAS No.: 254101-10-5 Molecular Formula: C17H23NO6 Formula Weight: 337.37 Beilstein Reference: 8430789 MDL No.: MFCD01862861

Formula: C₁₇H₂₃NO₆
Storage Temperature: Ambient
MDL Number: MFCD01862861
CAS Number: 254101-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR