Fmoc-L-β-homoaspartic acid-gamma-t-butyl ester 95%

Supplier: Thermo Scientific Chemicals

H52190-03 H52190-MD
AAH52190-03EA 406 USD
AAH52190-03 AAH52190-MD
Fmoc-L-β-homoaspartic acid-gamma-t-butyl ester 95%
Fmoc-L-beta-homoaspartic acid-gamma-t-butyl ester

Purity: 95% CAS No.: 209252-17-5 Molecular Formula: C24H27NO6 Formula Weight: 425.48 Beilstein Reference: 7952342 MDL No.: MFCD01862860

Formula: C₂₄H₂₇NO₆
Storage Temperature: Ambient
MDL Number: MFCD01862860
CAS Number: 209252-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR