Fmoc-L-Nw-boc-β-homolysine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-β-HomoLys(Boc)-OH
H52189-03
H52189-MD
AAH52189-03EA
398
USD
AAH52189-03
AAH52189-MD
Fmoc-L-Nw-boc-β-homolysine 95%
Fmoc-L-Nw-boc-beta-homolysine
Purity: 95%
CAS No.: 203854-47-1
Molecular Formula: C27H34N2O6
Formula Weight: 482.58
Beilstein Reference: 8022293
MDL No.: MFCD01863054
Formula:
C₂₇H₃₄N₂O₆ MW: 482.58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03002713 CAS Number: 203854-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...