Fmoc-L-β-homoglutamic acid-d-t-butyl ester 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-β-HoGlu(OtBu)-OH, Fmoc-β-HomoGlu(OtBu)-OH
H52188-03
H52188-MD
AAH52188-03EA
418
USD
AAH52188-03
AAH52188-MD
Fmoc-L-β-homoglutamic acid-d-t-butyl ester 95%
Fmoc-L-beta-homoglutamic acid-d-t-butyl ester
Purity: 95%
CAS No.: 203854-49-3
Molecular Formula: C25H29NO6
Formula Weight: 439.51
Melting Point: 115-120°
Beilstein Reference: 8019724
MDL No.: MFCD01863053
Formula:
C₂₅H₂₉NO₆ MW: 439.51 g/mol Melting Pt: 115…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863053 CAS Number: 203854-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...