Fmoc-L-beta-homovaline 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-β-HoVal-OH, Fmoc-β-HomoVal-OH
H52182-03
H52182-MD
AAH52182-03EA
380
USD
AAH52182-03
AAH52182-MD
Fmoc-L-beta-homovaline 95%
Fmoc-L-beta-homovaline
Purity: 95%
CAS No.: 172695-33-9
Molecular Formula: C21H23NO4
Formula Weight: 353.42
Beilstein Reference: 7388393
MDL No.: MFCD01862853
Formula:
C₂₁H₂₃NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD01862853 CAS Number: 172695-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...