Fmoc-3-(2-naphthyl)-L-alanine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-2-Nal-OH
H52179-03
H52179-06
H52179-MD
AAH52179-03EA
129.92
USD
AAH52179-03
AAH52179-06
AAH52179-MD
Fmoc-3-(2-naphthyl)-L-alanine 95%
Fmoc-3-(2-naphthyl)-L-alanine
Purity: 95%
CAS No.: 112883-43-9
Molecular Formula: C28H23NO4
Formula Weight: 437.49
Beilstein Reference: 7668973
MDL No.: MFCD00144886
Formula:
C₂₈H₂₃NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00144886 CAS Number: 112883-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...