Fmoc-L-allylglycine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-Gly(allyl)-OH
H52177-03
H52177-06
H52177-MD
AAH52177-03EA
151
USD
AAH52177-03
AAH52177-06
AAH52177-MD
Fmoc-L-allylglycine 95%
Fmoc-L-allylglycine
Purity: 95%
CAS No.: 146549-21-5
Molecular Formula: C20H19NO4
Formula Weight: 337.37
Physical Form: Crystalline solid
Appearance: White
Beilstein Reference: 5884047
MDL No.: MFCD01311749
Formula:
C₂₀H₁₉NO₄ MW: 337.38 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01311749 CAS Number: 146549-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...