Boc-L-β-homoleucine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-β-HoLeu-OH, (S)-3-((tert-Butoxycarbonyl)amino)-5-methylhexanoic acid

H52173-03 H52173-MD
AAH52173-03EA 365 USD
AAH52173-03 AAH52173-MD
Boc-L-β-homoleucine 95%
Boc-L-beta-homoleucine

Purity: 95% CAS No.: 132549-43-0 Molecular Formula: C12H23NO4 Formula Weight: 245.32 Melting Point: 53-55° Beilstein Reference: 4251252 MDL No.: MFCD02101665

Formula: C₁₂H₂₃NO₄
MW: 245.32 g/mol
Melting Pt: 53…55 °C
Storage Temperature: Ambient
MDL Number: MFCD02101665
CAS Number: 132549-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR