Fmoc-L-4-pyridylalanine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Fmoc-3-(4-pyridyl)-L-alanine, Fmoc-3-(4-pyridyl)-Ala-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(4-pyridyl)-L-alanine, Fmoc-4-Pal-OH
H52172-03
H52172-06
H52172-MD
AAH52172-03EA
212
USD
AAH52172-03
AAH52172-06
AAH52172-MD
Fmoc-L-4-pyridylalanine 95%
Fmoc-L-4-pyridylalanine
Purity: 95%
CAS No.: 169555-95-7
Molecular Formula: C23H20N2O4
Formula Weight: 388.42
Beilstein Reference: 7316905
MDL No.: MFCD00672566
Formula:
C₂₃H₂₀N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00672566 CAS Number: 169555-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...